Search
Search

Cyth4| Gene Symbol: Cyth4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028195.3 | NP_082471.2 | 10 - 11 | 1042 | 95 | |
| GenBank mRNA | AF079972.1 | - | 10 - 11 | 973 | 95 | |
| AK152306.1 | - | 10 - 11 | 948 | 95 | ||
| AK152909.1 | - | 10 - 11 | 948 | 95 | ||
| AK157026.1 | - | 10 - 11 | 948 | 95 | ||
| AK220198.1 | - | 10 - 11 | 971 | 95 | ||
| BC018505.1 | - | 9 - 10 | 866 | 95 | ||
| BC139054.1 | - | 10 - 11 | 1015 | 8862076 | 95 | |
| BC139056.1 | - | 10 - 11 | 1015 | 8862078 | 95 | |
| BC145528.1 | - | 10 - 11 | 1042 | 95 | ||
Set Membership: |
|