Search
Search

Tnrc18| Gene Symbol: Tnrc18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122730.2 | NP_001116202.2 | 2 - 3 | 551 | 75 | |
| NM_178242.2 | NP_839973.1 | 2 - 3 | 551 | 75 | ||
| GenBank mRNA | BC049818.1 | - | 2 - 3 | 551 | 6511384 | 75 |
Set Membership: |
|