Search
Search

Kank1| Gene Symbol: Kank1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181404.5 | NP_852069.4 | 3 - 4 | 328 | 99 | |
| GenBank mRNA | AK020911.1 | - | 3 - 4 | 556 | 99 | |
| AK041011.1 | - | 3 - 4 | 328 | 99 | ||
Set Membership: |
|