Search
Search

Cars2| Gene Symbol: Cars2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024248.1 | NP_077210.1 | 4 - 5 | 514 | 102 | |
| GenBank mRNA | AK009937.1 | - | 4 - 5 | 476 | 102 | |
| AK038968.1 | - | 4 - 5 | 527 | 102 | ||
| AK041962.1 | - | 4 - 5 | 488 | 102 | ||
| AK054486.1 | - | 4 - 5 | 511 | 102 | ||
| CX204497.1 | - | 4 - 5 | 515 | 102 | ||
Set Membership: |
|