Search
Search
| Gene Symbol: Tpte | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199257.2 | NP_954866.2 | 25 - 26 | 2104 | 85 | |
| GenBank mRNA | AJ311311.1 | - | 25 - 26 | 2107 | 85 | |
| AJ311312.1 | - | 25 - 26 | 2192 | 85 | ||
| BC107329.2 | - | 25 - 26 | 1899 | 40054415 | 85 | |
Set Membership: |
|