Search
Search

Tapt1| Gene Symbol: Tapt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173764.3 | NP_776125.2 | 9 - 10 | 1100 | 106 | |
| GenBank mRNA | AK030002.1 | - | 4 - 5 | 342 | 106 | |
| BC096038.1 | - | 6 - 7 | 579 | 106 | ||
| BC110670.1 | - | 8 - 9 | 821 | 106 | ||
Set Membership: |
|