Search
Search
| Gene Symbol: Ccnc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122982.2 | NP_001116454.1 | 3 - 4 | 402 | 95 | |
| NM_001290420.1 | NP_001277349.1 | 3 - 4 | 402 | 95 | ||
| NM_001290422.1 | NP_001277351.1 | 3 - 4 | 402 | 95 | ||
| NM_016746.4 | NP_058026.2 | 3 - 4 | 402 | 95 | ||
| GenBank mRNA | AK009615.1 | - | 3 - 4 | 347 | 95 | |
| AK035728.1 | - | 3 - 4 | 347 | 95 | ||
| AK035762.1 | - | 3 - 4 | 363 | 95 | ||
| AK037957.1 | - | 3 - 4 | 403 | 95 | ||
| AK135472.1 | - | 3 - 4 | 398 | 95 | ||
| AK153035.1 | - | 3 - 4 | 335 | 95 | ||
| BB631264.1 | - | 3 - 4 | 400 | 95 | ||
| BC003344.1 | - | 3 - 4 | 335 | 3257730 | 95 | |
| BC029211.1 | - | 3 - 4 | 348 | 4526925 | 95 | |
| BC037689.1 | - | 3 - 4 | 330 | 95 | ||
| BC062376.1 | - | 3 - 4 | 317 | 95 | ||
| BC087544.1 | - | 3 - 4 | 334 | 95 | ||
| BC100396.1 | - | 3 - 4 | 376 | 95 | ||
| BC120649.1 | - | 3 - 4 | 329 | 40129572 | 95 | |
| BC120677.1 | - | 3 - 4 | 329 | 40129600 | 95 | |
Set Membership: |
|