Search
Search

Tbc1d1| Gene Symbol: Tbc1d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289514.1 | NP_001276443.1 | 12 - 13 | 1977 | 64 | |
| NM_019636.3 | NP_062610.2 | 13 - 14 | 2558 | 64 | ||
| GenBank mRNA | AK122445.1 | - | 15 - 16 | 3646 | 64 | |
| AK135665.1 | - | 12 - 13 | 2149 | 64 | ||
| AK148356.1 | - | 12 - 13 | 1934 | 64 | ||
| AK154591.1 | - | 14 - 15 | 2594 | 64 | ||
| AK159256.1 | - | 13 - 14 | 2554 | 64 | ||
| AK164923.1 | - | 9 - 10 | 1262 | 64 | ||
| AK168763.1 | - | 2 - 3 | 263 | 64 | ||
| BC004675.1 | - | 2 - 3 | 216 | 64 | ||
| BC059824.1 | - | 13 - 14 | 2732 | 64 | ||
| BC117931.1 | - | 12 - 13 | 2226 | 40097712 | 64 | |
| U33005.1 | - | 13 - 14 | 2514 | 64 | ||
Set Membership: |
|