Search
Search

Aftph| Gene Symbol: Aftph | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252503.2 | NP_001239432.1 | 2 - 3 | 2208 | 61 | |
| NM_001290545.1 | NP_001277474.1 | 2 - 3 | 2208 | 61 | ||
| NM_181411.4 | NP_852076.1 | 2 - 3 | 2208 | 61 | ||
| GenBank mRNA | AK029681.1 | - | 2 - 3 | 2107 | 61 | |
| AK030572.1 | - | 2 - 3 | 2152 | 61 | ||
| AK047225.1 | - | 1 - 2 | 1272 | 61 | ||
| AK136815.1 | - | 1 - 2 | 1460 | 61 | ||
| AK138793.1 | - | 2 - 3 | 2130 | 61 | ||
| BC004630.1 | - | 1 - 2 | 497 | 61 | ||
| BC052036.1 | - | 2 - 3 | 2136 | 5702534 | 61 | |
Set Membership: |
|