Search
Search
| Gene Symbol: Hmmr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013552.2 | NP_038580.2 | 15 - 16 | 2116 | 159 | |
| GenBank mRNA | AF031932.1 | - | 15 - 16 | 2101 | 159 | |
| AF079222.1 | - | 16 - 17 | 2065 | 159 | ||
| X64550.1 | - | 11 - 12 | 1729 | 159 | ||
Set Membership: |
|