Search
Search

Tex16| Gene Symbol: Tex16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031382.2 | NP_113559.2 | 2 - 3 | 949 | 143 | |
| GenBank mRNA | AF285573.1 | - | 2 - 3 | 949 | 143 | |
| BC132332.1 | - | 2 - 3 | 973 | 40130609 | 143 | |
Set Membership: |
|