Search
Search

Cep95| Gene Symbol: Cep95 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166685.1 | NP_001160157.1 | 1 - 2 | 126 | 94 | |
| NM_177088.3 | NP_796062.2 | 1 - 2 | 126 | 94 | ||
| GenBank mRNA | AK052705.1 | - | 1 - 2 | 54 | 94 | |
| AK076400.1 | - | 1 - 2 | 104 | 94 | ||
| AK165351.1 | - | 1 - 2 | 95 | 94 | ||
| BC090675.1 | - | 1 - 2 | 84 | 94 | ||
| BC116308.1 | - | 1 - 2 | 92 | 40095242 | 94 | |
| BC116309.1 | - | 1 - 2 | 92 | 94 | ||
Set Membership: |
|