Search
Search

Smarcd2| Gene Symbol: Smarcd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130187.1 | NP_001123659.1 | 3 - 4 | 704 | 63 | |
| NM_031878.2 | NP_114084.2 | 2 - 3 | 338 | 63 | ||
| GenBank mRNA | AK159259.1 | - | 3 - 4 | 541 | 63 | |
| AK166123.1 | - | 3 - 4 | 497 | 63 | ||
| BC005732.1 | - | 2 - 3 | 328 | 2647494 | 63 | |
Set Membership: |
|