Search
Search

Nfat5| Gene Symbol: Nfat5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286260.1 | NP_001273189.1 | 7 - 8 | 1803 | 129 | |
| NM_018823.2 | NP_061293.2 | 8 - 9 | 1857 | 129 | ||
| NM_133957.3 | NP_598718.2 | 9 - 10 | 1919 | 129 | ||
| GenBank mRNA | AF162853.1 | - | 7 - 8 | 1714 | 129 | |
| AF200687.1 | - | 8 - 9 | 1492 | 129 | ||
| AF228706.1 | - | 5 - 6 | 1557 | 129 | ||
| AF369980.1 | - | 9 - 10 | 1558 | 129 | ||
| AF453571.2 | - | 9 - 10 | 1527 | 129 | ||
| AK134022.1 | - | 9 - 10 | 1923 | 129 | ||
Set Membership: |
|