Search
Search
| Gene Symbol: Ggt7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144786.2 | NP_659035.2 | 12 - 13 | 1636 | 58 | |
| GenBank mRNA | AK032051.1 | - | 12 - 13 | 1634 | 58 | |
| AK131175.1 | - | 9 - 10 | 3659 | 58 | ||
| BC005772.1 | - | 12 - 13 | 1617 | 3602314 | 58 | |
Set Membership: |
|