Search
Search

Spire2| Gene Symbol: Spire2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172287.2 | NP_758491.1 | 8 - 9 | 1334 | 65 | |
| GenBank mRNA | BC026502.1 | - | 7 - 8 | 1162 | 4506974 | 65 |
| BC049152.1 | - | 8 - 9 | 1334 | 6311485 | 65 | |
| BC054072.1 | - | 8 - 9 | 1334 | 65 | ||
Set Membership: |
|