Search
Search

Hsd17b2| Gene Symbol: Hsd17b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008290.2 | NP_032316.2 | 3 - 4 | 718 | 68 | |
| GenBank mRNA | AK004950.1 | - | 3 - 4 | 720 | 68 | |
| BE649625.1 | - | 2 - 3 | 349 | 68 | ||
| X95685.1 | - | 3 - 4 | 679 | 68 | ||
| Y09517.1 | - | 3 - 4 | 762 | 68 | ||
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | AI194967.1 | - | Chr. - - [-] on Build | 68 | |||
Set Membership: |
|