Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Camkk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018883.2 | NP_061371.2 | 6 - 7 | 809 | 93 | |
GenBank mRNA | AK144483.1 | - | 6 - 7 | 810 | 93 | |
BC017529.1 | - | 6 - 7 | 774 | 4924656 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Apoptosis Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |