Search
Search

Glb1l| Gene Symbol: Glb1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029010.1 | NP_083286.1 | 12 - 13 | 1375 | 113 | |
| GenBank mRNA | AK014667.1 | - | 10 - 11 | 1398 | 113 | |
| AK029925.1 | - | 12 - 13 | 1375 | 113 | ||
| BC021773.1 | - | 11 - 12 | 1128 | 4222994 | 113 | |
Set Membership: |
|