| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0400025246.mm.1 |
NM_027612.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400025246.mm.1 |
NM_027612.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400025246.mm.1 |
TC0400003174.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400025247.mm.1 |
NM_001163494.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400025247.mm.1 |
NM_001163494.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400025247.mm.1 |
NM_027612.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400025247.mm.1 |
NM_027612.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400025247.mm.1 |
TC0400003174.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400025248.mm.1 |
NM_001163494.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400025248.mm.1 |
NM_001163494.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400025248.mm.1 |
NM_027612.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400025248.mm.1 |
NM_027612.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400025248.mm.1 |
TC0400003174.mm.1 |
MTA TC |