Search
Search

4921539E11Rik| Gene Symbol: 4921539E11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027612.1 | NP_081888.1 | 2 - 3 | 659 | 123 | |
| GenBank mRNA | AK015018.1 | - | 2 - 3 | 654 | 123 | |
| AK161421.1 | - | 2 - 3 | 659 | 123 | ||
| BC107405.1 | - | 2 - 3 | 600 | 40058778 | 123 | |
Set Membership: |
|