Search
Search

Fbxo15| Gene Symbol: Fbxo15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015798.3 | NP_056613.1 | 4 - 5 | 612 | 81 | |
| GenBank mRNA | AF176530.1 | - | 4 - 5 | 596 | 81 | |
| AK082809.1 | - | 4 - 5 | 633 | 81 | ||
| AK166591.1 | - | 4 - 5 | 613 | 81 | ||
| AK166599.1 | - | 4 - 5 | 614 | 81 | ||
| AK166918.1 | - | 4 - 5 | 609 | 81 | ||
| AK167118.1 | - | 4 - 5 | 613 | 81 | ||
| AK167270.1 | - | 4 - 5 | 612 | 81 | ||
Set Membership: |
|