| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0700014378.mm.1 |
NM_212441.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700014378.mm.1 |
NM_212441.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700014378.mm.1 |
TC0700001715.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700014376.mm.1 |
NM_016870.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700014376.mm.1 |
NM_016870.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700014376.mm.1 |
NM_212441.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700014376.mm.1 |
NM_212441.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700014376.mm.1 |
NM_212442.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700014376.mm.1 |
NM_212442.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700014376.mm.1 |
TC0700001715.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700014381.mm.1 |
NM_016870.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700014381.mm.1 |
NM_016870.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700014381.mm.1 |
NM_212441.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700014381.mm.1 |
NM_212441.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700014381.mm.1 |
NM_212442.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700014381.mm.1 |
NM_212442.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700014381.mm.1 |
TC0700001715.mm.1 |
MTA TC |