Search
Search

Zcchc10| Gene Symbol: Zcchc10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026479.4 | NP_080755.2 | 1 - 2 | 56 | 84 | |
| GenBank mRNA | AK010803.1 | - | 1 - 2 | 67 | 84 | |
| AK020177.1 | - | 1 - 2 | 57 | 84 | ||
| AK027985.1 | - | 1 - 2 | 64 | 84 | ||
| AK169856.1 | - | 1 - 2 | 57 | 84 | ||
| BC025078.1 | - | 1 - 2 | 72 | 4504126 | 84 | |
| BC087902.1 | - | 1 - 2 | 56 | 30505854 | 84 | |
Set Membership: |
|