Search
Search
| Gene Symbol: Sctr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012322.2 | NP_001012322.2 | 11 - 12 | 1337 | 79 | |
| GenBank mRNA | AK032652.1 | - | 11 - 12 | 1337 | 79 | |
| AY255623.1 | - | 3 - 4 | 226 | 79 | ||
| BC089355.1 | - | 10 - 11 | 1238 | 30551620 | 79 | |
Set Membership: |
|