Search
Search

Dis3l2| Gene Symbol: Dis3l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172157.1 | NP_001165628.1 | 5 - 6 | 528 | 68 | |
| GenBank mRNA | AK031180.1 | - | 5 - 6 | 511 | 68 | |
| AK040613.1 | - | 5 - 6 | 505 | 68 | ||
| AK184309.1 | - | 1 - 2 | 102 | 68 | ||
| AK214974.1 | - | 1 - 2 | 102 | 68 | ||
Set Membership: |
|