Search
Search

Espl1| Gene Symbol: Espl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014976.2 | NP_001014976.1 | 28 - 29 | 6030 | 68 | |
| GenBank mRNA | AK129072.1 | - | 28 - 29 | 5997 | 68 | |
| AY588413.1 | - | 28 - 29 | 5968 | 68 | ||
| BC082603.1 | - | 28 - 29 | 6030 | 30668743 | 68 | |
| BC145844.1 | - | 28 - 29 | 6001 | 40131147 | 68 | |
| BC145846.1 | - | 28 - 29 | 5947 | 40131149 | 68 | |
Set Membership: |
|