Search
Search

Espl1| Gene Symbol: Espl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014976.2 | NP_001014976.1 | 8 - 9 | 2062 | 72 | |
| GenBank mRNA | AK129072.1 | - | 8 - 9 | 2029 | 72 | |
| AY588413.1 | - | 8 - 9 | 2000 | 72 | ||
| BC082603.1 | - | 8 - 9 | 2062 | 30668743 | 72 | |
| BC145844.1 | - | 8 - 9 | 2033 | 40131147 | 72 | |
| BC145846.1 | - | 8 - 9 | 1979 | 40131149 | 72 | |
Set Membership: |
|