Search
Search

Adamts6| Gene Symbol: Adamts6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081020.1 | NP_001074489.1 | 17 - 18 | 2601 | 78 | |
| GenBank mRNA | AK030315.1 | - | 17 - 18 | 2788 | 78 | |
| BC172116.1 | - | 17 - 18 | 2538 | 78 | ||
Set Membership: |
|