Search
Search

Mtmr12| Gene Symbol: Mtmr12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172958.3 | NP_766546.1 | 7 - 8 | 816 | 110 | |
| GenBank mRNA | AK030139.1 | - | 7 - 8 | 883 | 110 | |
| AK050099.1 | - | 7 - 8 | 809 | 110 | ||
| AK050236.1 | - | 7 - 8 | 779 | 110 | ||
| AK122539.1 | - | 7 - 8 | 874 | 110 | ||
| BC042573.1 | - | 7 - 8 | 805 | 110 | ||
| BC054352.1 | - | 7 - 8 | 816 | 3661020 | 110 | |
Set Membership: |
|