Search
Search
| Gene Symbol: Eed | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021876.3 | NP_068676.1 | 8 - 9 | 1317 | 149 | |
| GenBank mRNA | AK077664.1 | - | 8 - 9 | 902 | 149 | |
| AK131976.1 | - | 8 - 9 | 884 | 149 | ||
| AK133890.1 | - | 8 - 9 | 878 | 149 | ||
| BC012966.1 | - | 8 - 9 | 1316 | 3991086 | 149 | |
| U78103.1 | - | 8 - 9 | 1319 | 149 | ||
| U97675.1 | - | 8 - 9 | 1324 | 149 | ||
Set Membership: |
|