Search
Search

Klhl29| Gene Symbol: Klhl29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164493.1 | NP_001157965.1 | 5 - 6 | 1668 | 93 | |
| GenBank mRNA | AK122572.1 | - | 5 - 6 | 1321 | 93 | |
| BC065168.1 | - | 5 - 6 | 1668 | 93 | ||
| BC138283.1 | - | 4 - 5 | 1024 | 8861302 | 93 | |
| BC145230.1 | - | 4 - 5 | 1024 | 93 | ||
| BC145748.1 | - | 4 - 5 | 1024 | 40131051 | 93 | |
Set Membership: |
|