Search
Search
| Gene Symbol: Ctbs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293672.1 | NP_001280601.1 | 2 - 3 | 296 | 99 | |
| NM_028836.4 | NP_083112.1 | 2 - 3 | 296 | 99 | ||
| GenBank mRNA | AK008791.2 | - | 2 - 3 | 271 | 99 | |
| BC022594.1 | - | 1 - 2 | 57 | 99 | ||
| DQ100384.1 | - | 2 - 3 | 270 | 99 | ||
Set Membership: |
|