Search
Search

Abca4| Gene Symbol: Abca4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007378.1 | NP_031404.1 | 34 - 35 | 4927 | 88 | |
| GenBank mRNA | AF000149.1 | - | 34 - 35 | 4927 | 88 | |
| AK052916.1 | - | 11 - 12 | 1285 | 88 | ||
| BC057853.1 | - | 34 - 35 | 4927 | 6492139 | 88 | |
Set Membership: |
|