Search
Search

Akr1c19| Gene Symbol: Akr1c19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013785.3 | NP_001013807.2 | 6 - 7 | 714 | 88 | |
| GenBank mRNA | BC087964.1 | - | 6 - 7 | 714 | 6767816 | 88 |
Set Membership: |
|