Search
Search

Il1f6| Gene Symbol: Il1f6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019450.3 | NP_062323.1 | 3 - 4 | 287 | 64 | |
| GenBank mRNA | AF200493.1 | - | 2 - 3 | 126 | 64 | |
| AF206697.1 | - | 3 - 4 | 201 | 64 | ||
| AK004061.1 | - | 3 - 4 | 287 | 64 | ||
| AK224931.1 | - | 3 - 4 | 221 | 64 | ||
| AK224932.1 | - | 3 - 4 | 234 | 64 | ||
| BC117102.1 | - | 2 - 3 | 160 | 40126421 | 64 | |
| BC117104.1 | - | 2 - 3 | 160 | 40126423 | 64 | |
Set Membership: |
|