Search
Search

Fetub| Gene Symbol: Fetub | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083904.1 | NP_001077373.1 | 4 - 5 | 634 | 120 | |
| NM_001083905.1 | NP_001077374.1 | 5 - 6 | 804 | 120 | ||
| NM_021564.2 | NP_067539.1 | 5 - 6 | 869 | 120 | ||
| GenBank mRNA | AJ242927.1 | - | 5 - 6 | 790 | 120 | |
| AK009310.1 | - | 5 - 6 | 859 | 120 | ||
| AK009554.1 | - | 5 - 6 | 803 | 120 | ||
| AK037043.1 | - | 5 - 6 | 805 | 120 | ||
| AY138459.1 | - | 5 - 6 | 854 | 120 | ||
| AY138460.1 | - | 4 - 5 | 404 | 120 | ||
| BC018341.1 | - | 5 - 6 | 734 | 4194184 | 120 | |
Set Membership: |
|