Search
Search

Thap7| Gene Symbol: Thap7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026909.2 | NP_081185.1 | 1 - 2 | 30 | 78 | |
| GenBank mRNA | AK154641.1 | - | 1 - 2 | 116 | 78 | |
| BC018221.1 | - | 1 - 2 | 30 | 4215606 | 78 | |
Set Membership: |
|