Search
Search

Diap2| Gene Symbol: Diap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172493.2 | NP_766081.1 | 25 - 26 | 3381 | 80 | |
| GenBank mRNA | AB713187.1 | - | 26 - 27 | 3176 | 80 | |
| AK083254.1 | - | 18 - 19 | 2313 | 80 | ||
| AK088648.1 | - | 25 - 26 | 3315 | 80 | ||
| AY312280.1 | - | 25 - 26 | 3318 | 80 | ||
| BB430043.2 | - | 2 - 3 | 263 | 80 | ||
| BC100759.1 | - | 2 - 3 | 428 | 80 | ||
| Y15910.1 | - | 6 - 7 | 888 | 80 | ||
Set Membership: |
|