Search
Search

Pdcd6| Gene Symbol: Pdcd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011051.3 | NP_035181.1 | 4 - 5 | 486 | 63 | |
| GenBank mRNA | AK008610.1 | - | 4 - 5 | 410 | 63 | |
| AK028223.1 | - | 4 - 5 | 441 | 63 | ||
| AK078044.1 | - | 4 - 5 | 414 | 63 | ||
| AK150335.1 | - | 4 - 5 | 404 | 63 | ||
| BC040079.1 | - | 4 - 5 | 437 | 3156800 | 63 | |
| BU604816.1 | - | 4 - 5 | 489 | 63 | ||
| U49112.1 | - | 4 - 5 | 423 | 63 | ||
| X14938.1 | - | 3 - 4 | 253 | 63 | ||
Set Membership: |
|