| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0200018424.mm.1 |
NM_001080811.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200018424.mm.1 |
NM_001080811.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200018424.mm.1 |
TC0200002331.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200018421.mm.1 |
NM_001080811.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200018421.mm.1 |
NM_001080811.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200018421.mm.1 |
TC0200002331.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200018422.mm.1 |
NM_001080811.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200018422.mm.1 |
NM_001080811.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200018422.mm.1 |
TC0200002331.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200018425.mm.1 |
NM_001080811.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200018425.mm.1 |
NM_001080811.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200018425.mm.1 |
TC0200002331.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200018423.mm.1 |
NM_001080811.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200018423.mm.1 |
NM_001080811.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200018423.mm.1 |
TC0200002331.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200018426.mm.1 |
NM_001080811.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200018426.mm.1 |
NM_001080811.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200018426.mm.1 |
TC0200002331.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200018427.mm.1 |
NM_001080811.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200018427.mm.1 |
NM_001080811.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200018427.mm.1 |
TC0200002331.mm.1 |
MTA TC |