Search
Search

Sec14l5| Gene Symbol: Sec14l5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127725.1 | NP_001121197.1 | 3 - 4 | 276 | 92 | |
| GenBank mRNA | BC157907.1 | - | 3 - 4 | 342 | 9088109 | 92 |
| BC157990.1 | - | 3 - 4 | 276 | 9007304 | 92 | |
Set Membership: |
|