Search
Search

Fbxl17| Gene Symbol: Fbxl17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015794.1 | NP_056609.1 | 9 | 9277 | 97 | |
| GenBank mRNA | AK018254.1 | - | 1 | 321 | 97 | |
| AK086279.1 | - | 1 | 1153 | 97 | ||
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ephrin A5 | Efna5 |
AL-1, AV158822, EFL-5, Ephrin-A5, Epl7, LERK-7, RAGS |
-171.0kb | Sense | Chr. 17 - 62602957 - 62881317 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 4930405O22 gene | 4930405O22Rik | 250.6kb | Sense | Chr. 17 - 63305446 - 63312973 on NCBI Build GRCm38 |
Set Membership: |
|