Search
Search

Lrrc8d| Gene Symbol: Lrrc8d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK013401.1 | - | 1 | 647 | 89 | |
| AK170452.1 | - | 2 | 2822 | 89 | ||
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 42166 |
Gm42166 | -26.6kb | Anti-sense | Chr. 5 - 105697029 - 105704089 on NCBI Build GRCm38 | |
| Downstream | RIKEN cDNA D830014E11 gene | D830014E11Rik | 95.6kb | Sense | Chr. 5 - 105827095 - 105832437 on NCBI Build GRCm38 |
Set Membership: |
|