Search
Search

Cilp2| Gene Symbol: Cilp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK004006.1 | - | 1 | 52 | 74 | |
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | pre B cell leukemia homeobox 4 | Pbx4 |
2410015M21Rik, AI429113, AW045266 |
-8.1kb | Anti-sense | Chr. 8 - 69832476 - 69872292 on NCBI Build GRCm38 |
| Downstream |
NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 13 |
Ndufa13 |
2700054G14Rik, AU022060, CDA016, CGI-39, GRIM-19, Grim19 |
6.8kb | Sense | Chr. 8 - 69887788 - 69902558 on NCBI Build GRCm38 |
Set Membership: |
|