Search
Search

Gpr21| Gene Symbol: Gpr21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177383.4 | NP_796357.1 | 2 | 1247 | 83 | |
| GenBank mRNA | AK048682.1 | - | 2 | 1248 | 83 | |
| AK139361.1 | - | 2 | 1249 | 83 | ||
| AK141999.1 | - | 1281 | 83 | |||
| BC112377.1 | - | 1 | 1130 | 40057664 | 83 | |
Set Membership: |
|