Search
Search

Kcnj16| Gene Symbol: Kcnj16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010604.3 | NP_034734.3 | 1 - 2 | 59 | 105 | |
| GenBank mRNA | AK078905.1 | - | 1 - 2 | 60 | 105 | |
| AK128942.1 | - | 1 - 2 | 59 | 105 | ||
Set Membership: |
|