Search
Search

H2-M10.1| Gene Symbol: H2-M10.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013544.3 | NP_038572.3 | 2 | 254 | 81 | |
| GenBank mRNA | AF539597.1 | - | 2 | 254 | 81 | |
| AY211079.1 | - | 2 | 254 | 81 | ||
| BC104320.1 | - | 2 | 120 | 81 | ||
| BC104321.1 | - | 2 | 246 | 81 | ||
| BC119458.2 | - | 2 | 254 | 40044798 | 81 | |
| BC152395.1 | - | 2 | 246 | 81 | ||
Set Membership: |
|