Search
Search
| Gene Symbol: Dok6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039173.1 | NP_001034262.1 | 2 - 3 | 177 | 109 | |
| GenBank mRNA | AB221044.1 | - | 2 - 3 | 177 | 109 | |
| BC138955.1 | - | 2 - 3 | 228 | 8861977 | 109 | |
| BC138957.1 | - | 2 - 3 | 228 | 8861979 | 109 | |
Set Membership: |
|