Search
Search

HMG20A| Gene Symbol: HMG20A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004017834.4 | XP_004017883.1 | 9 - 10 | 1364 | 68 | |
| XM_012098660.3 | XP_011954050.1 | 9 - 10 | 1099 | 68 | ||
Set Membership: |
|